Product Details

SNP ID
rs115169407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30679258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCTTCCTCATCCTCTTCGTC[G/T]TCGGGTTGGGCTGCTGGAGGGTTGG
Phenotype
MIM: 603405 MIM: 610990
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DHX16 PubMed Links

Gene Details

Gene
DHX16
Gene Name
DEAH-box helicase 16
There are no transcripts associated with this gene.

Gene
NRM
Gene Name
nurim (nuclear envelope membrane protein)
There are no transcripts associated with this gene.

Gene
PPP1R18
Gene Name
protein phosphatase 1 regulatory subunit 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134870.1 1974 Missense Mutation GAA,GAC E581D NP_001128342.1
NM_133471.3 1974 Missense Mutation GAA,GAC E581D NP_597728.1

View Full Product Details