Product Details

SNP ID
rs111323174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32191886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTCAGTCTGCAGGGAGAGCAG[A/G]GAGGCCGAGCGGGGTCCCAGGGCTG
Phenotype
MIM: 600214 MIM: 164951 MIM: 176311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGER PubMed Links

Gene Details

Gene
AGER
Gene Name
advanced glycosylation end product-specific receptor
There are no transcripts associated with this gene.

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276501.1 541 Silent Mutation TCC,TCT S56S NP_001263430.1
NM_022107.2 541 Silent Mutation TCC,TCT S56S NP_071390.1
Gene
NOTCH4
Gene Name
notch 4
There are no transcripts associated with this gene.

Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

View Full Product Details