Product Details

SNP ID
rs113543625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34287717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGATGATACCCAGGTCAGCAC[A/G]GTGTAAGGCATATATGGACACAAAG
Phenotype
MIM: 609228
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102723521 PubMed Links

Gene Details

Gene
LOC102723521
Gene Name
uncharacterized LOC102723521
There are no transcripts associated with this gene.

Gene
NUDT3
Gene Name
nudix hydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006703.3 2006 UTR 3 NP_006694.1
Gene
RPS10-NUDT3
Gene Name
RPS10-NUDT3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202470.2 2006 UTR 3 NP_001189399.1

View Full Product Details