Product Details

SNP ID
rs112222376
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35797235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAATGATCCCCCGGGGGCCAGG[A/G]GCTGCATAGGCCACAGAGAGGGCGA
Phenotype
MIM: 120105 MIM: 609427
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLPS PubMed Links

Gene Details

Gene
CLPS
Gene Name
colipase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252597.1 110 UTR 5 NP_001239526.1
NM_001252598.1 110 Silent Mutation GCC,GCT A18A NP_001239527.1
NM_001832.3 110 Silent Mutation GCC,GCT A18A NP_001823.1
Gene
CLPSL1
Gene Name
colipase like 1
There are no transcripts associated with this gene.

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
There are no transcripts associated with this gene.

View Full Product Details