Product Details

SNP ID
rs115639569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36400252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTGATGCTGCCAGTCCACAGA[C/T]CACAGTCTGAGTAGCAAGTATGAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PXT1 PubMed Links

Gene Details

Gene
PXT1
Gene Name
peroxisomal, testis specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152990.3 Intron NP_694535.2
XM_011514400.2 Intron XP_011512702.1

View Full Product Details