Product Details

SNP ID
rs112128222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:40378692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTACTCGCCCATTCTGCTCTT[C/G]TTCACCTCCGTATTTTCTCTCTCAC
Phenotype
MIM: 615676
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TDRG1 PubMed Links

Gene Details

Gene
TDRG1
Gene Name
testis development related 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details