Product Details

SNP ID
rs116667889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41053413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCAGAATGGGGAGGATCTGGA[A/G]AACCTGGACGACCCTGAGAAGCTGA
Phenotype
MIM: 604797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOBEC2 PubMed Links

Gene Details

Gene
APOBEC2
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006789.3 213 Silent Mutation GAA,GAG E22E NP_006780.1
Gene
TSPO2
Gene Name
translocator protein 2
There are no transcripts associated with this gene.

View Full Product Details