Product Details

SNP ID
rs115208681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41190269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGAGGGACAGTGACTTGCCAA[G/T]TTCACACTGAGATGAGTCCACCCTG
Phenotype
MIM: 609715
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TREML2 PubMed Links

Gene Details

Gene
TREML2
Gene Name
triggering receptor expressed on myeloid cells like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024807.3 3028 UTR 3 NP_079083.2
XM_011514917.2 3028 UTR 3 XP_011513219.1

View Full Product Details