Product Details

SNP ID
rs115599508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42227367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCGGTTTCACAGTACTTCCAA[A/G]TGGCACAAAGTAATTTCAGAACTCC
Phenotype
MIM: 611976 MIM: 610322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS10 PubMed Links

Gene Details

Gene
MRPS10
Gene Name
mitochondrial ribosomal protein S10
There are no transcripts associated with this gene.

Gene
TRERF1
Gene Name
transcriptional regulating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297573.1 5628 UTR 3 NP_001284502.1
NM_033502.3 5628 UTR 3 NP_277037.1
XM_011514741.2 5628 UTR 3 XP_011513043.1
XM_011514742.2 5628 UTR 3 XP_011513044.1
XM_011514743.2 5628 UTR 3 XP_011513045.1
XM_011514744.1 5628 UTR 3 XP_011513046.1
XM_011514745.1 5628 UTR 3 XP_011513047.1
XM_017011034.1 5628 UTR 3 XP_016866523.1
XM_017011035.1 5628 UTR 3 XP_016866524.1
XM_017011036.1 5628 UTR 3 XP_016866525.1
XM_017011037.1 5628 UTR 3 XP_016866526.1
XM_017011038.1 5628 UTR 3 XP_016866527.1
XM_017011039.1 5628 UTR 3 XP_016866528.1
XM_017011040.1 5628 UTR 3 XP_016866529.1
XM_017011041.1 5628 UTR 3 XP_016866530.1
XM_017011042.1 5628 UTR 3 XP_016866531.1
XM_017011043.1 5628 UTR 3 XP_016866532.1
XM_017011044.1 5628 UTR 3 XP_016866533.1
XM_017011045.1 5628 UTR 3 XP_016866534.1
XM_017011046.1 5628 UTR 3 XP_016866535.1
XM_017011047.1 5628 UTR 3 XP_016866536.1
XM_017011048.1 5628 UTR 3 XP_016866537.1
XM_017011049.1 5628 UTR 3 XP_016866538.1
XM_017011050.1 5628 UTR 3 XP_016866539.1
XM_017011051.1 5628 UTR 3 XP_016866540.1
XM_017011052.1 5628 UTR 3 XP_016866541.1
XM_017011053.1 5628 UTR 3 XP_016866542.1
XM_017011054.1 5628 UTR 3 XP_016866543.1
XM_017011055.1 5628 UTR 3 XP_016866544.1

View Full Product Details