Product Details

SNP ID
rs115433597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43331588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACATTCTAATCTTGTTCATCCT[C/T]TACAGGTACAACAATATACACATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF318 PubMed Links

Gene Details

Gene
ZNF318
Gene Name
zinc finger protein 318
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014345.2 3852 Intron NP_055160.2
XM_011514451.2 3852 Intron XP_011512753.1
XM_011514452.2 3852 UTR 3 XP_011512754.1

View Full Product Details