Product Details

SNP ID
rs114096520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49728802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCAACTGATGTTTACAGGTTAA[C/T]GTGAGCTTCAAACTTTTACAGTTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRISP3 PubMed Links

Gene Details

Gene
CRISP3
Gene Name
cysteine rich secretory protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190986.1 793 Silent Mutation ACA,ACG T245T NP_001177915.1
NM_006061.2 793 Silent Mutation ACA,ACG T235T NP_006052.2

View Full Product Details