Product Details

SNP ID
rs114514860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:8081898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTTTCTTACCATTTTAGTCCT[G/T]TCATCGCTCTTTCATTTCAGATAAT
Phenotype
MIM: 609206
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EEF1E1 PubMed Links

Gene Details

Gene
EEF1E1
Gene Name
eukaryotic translation elongation factor 1 epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135650.1 Intron NP_001129122.1
NM_004280.4 Intron NP_004271.1
Gene
EEF1E1-BLOC1S5
Gene Name
EEF1E1-BLOC1S5 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SCARNA27
Gene Name
small Cajal body-specific RNA 27
There are no transcripts associated with this gene.

View Full Product Details