Product Details
- SNP ID
-
rs114984145
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:9776965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATGCAGGTGCTTGGAGTGCTTC[A/G]GGAAAGCCACAGCTTTGATATGGGG
- Phenotype
-
MIM: 614287
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OFCC1
PubMed Links
Gene Details
- Gene
- OFCC1
- Gene Name
- orofacial cleft 1 candidate 1
There are no transcripts associated with this gene.
View Full Product Details