Product Details
- SNP ID
-
rs117969779
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44963615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTACCCCCGTCTTGTGGGAAAGA[C/G]AATTTATGGGAACAGCAGCTGCTAA
- Phenotype
-
MIM: 600642
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1G
PubMed Links
Gene Details
- Gene
- MYO1G
- Gene Name
- myosin IG
View Full Product Details