Product Details

SNP ID
rs113208188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6689846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAACACAGTGAATTCCTATGTTA[C/T]ACAGCATTGTGCTCTGCACTGTTAT
Phenotype
MIM: 194536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF12 PubMed Links

Gene Details

Gene
ZNF12
Gene Name
zinc finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006956.2 3536 UTR 3 NP_008887.2
NM_016265.3 3536 UTR 3 NP_057349.2

View Full Product Details