Product Details

SNP ID
rs116884794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:967333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTCTATCCAGTTACCTTTGTA[A/C]GTTAGAAAAGCAGGAGCTCAATACA
Phenotype
MIM: 610429
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COX19 PubMed Links

Gene Details

Gene
COX19
Gene Name
COX19, cytochrome c oxidase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031617.2 2408 UTR 3 NP_001026788.1

View Full Product Details