Product Details

SNP ID
rs116837570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149875992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGTTTTCAGGCTGCACCCTAC[A/G]CCCATGAAATTACAACTCTCGAGAT
Phenotype
MIM: 611019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR3C PubMed Links

Gene Details

Gene
ACTR3C
Gene Name
ARP3 actin-related protein 3 homolog C
There are no transcripts associated with this gene.

Gene
ATP6V0E2
Gene Name
ATPase H+ transporting V0 subunit e2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100592.2 Intron NP_001094062.1
NM_001289990.1 Intron NP_001276919.1
NM_145230.3 Intron NP_660265.2
Gene
ATP6V0E2-AS1
Gene Name
ATP6V0E2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF862
Gene Name
zinc finger protein 862
There are no transcripts associated with this gene.

View Full Product Details