Product Details
- SNP ID
-
rs104886484
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:151180965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGGGAGGGCCCCTGGCCAGACAC[G/A]GACGGCGCCATGGTTGAGCAGAGCC
- Phenotype
-
MIM: 615054
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ASB10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201566253] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASB10
- Gene Name
- ankyrin repeat and SOCS box containing 10
- Gene
- IQCA1L
- Gene Name
- IQ motif containing with AAA domain 1 like
There are no transcripts associated with this gene.
View Full Product Details