Product Details
- SNP ID
-
rs114619941
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:152051746 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTACTCTGCCTAAGCAAAAATAGG[A/G]AATAGAAAAACCTCTCTTTATGCTC
- Phenotype
-
MIM: 615130
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GALNT11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11291792] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GALNT11
- Gene Name
- polypeptide N-acetylgalactosaminyltransferase 11
View Full Product Details