Product Details

SNP ID
rs117817732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152761060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGTTTGCTACAGTCCATCTCCT[A/G]CTGTTAGTTCCTTCCTTTTGCTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR3B PubMed Links

Gene Details

Gene
ACTR3B
Gene Name
ARP3 actin related protein 3 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040135.2 Intron NP_001035225.1
NM_020445.5 Intron NP_065178.1
XM_006716065.2 Intron XP_006716128.1
XM_006716066.1 Intron XP_006716129.1
XM_006716067.3 Intron XP_006716130.1
XM_011516425.2 Intron XP_011514727.1
XM_011516426.2 Intron XP_011514728.1
XM_011516427.1 Intron XP_011514729.1
XM_017012460.1 Intron XP_016867949.1
XM_017012461.1 Intron XP_016867950.1
XM_017012462.1 Intron XP_016867951.1
XM_017012463.1 Intron XP_016867952.1
XM_017012464.1 Intron XP_016867953.1
XM_017012465.1 Intron XP_016867954.1

View Full Product Details