Product Details

SNP ID
rs116080696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73442541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGTAGTCCTCGGCCTCATCTCT[A/G]GTCACAGGCTCCCTGTGGAGAAGAA
Phenotype
MIM: 605681 MIM: 601766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAZ1B PubMed Links

Gene Details

Gene
BAZ1B
Gene Name
bromodomain adjacent to zinc finger domain 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032408.3 4472 Silent Mutation ACC,ACT T1369T NP_115784.1
XM_017012773.1 4472 Silent Mutation ACC,ACT T1369T XP_016868262.1
Gene
FZD9
Gene Name
frizzled class receptor 9
There are no transcripts associated with this gene.

View Full Product Details