Product Details

SNP ID
rs112869815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75882226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTAATGTCTGCGGGCTGTCCAT[C/T]GGGCTGGCCTGTATCCTTCCTAGCA
Phenotype
MIM: 615203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHBDD2 PubMed Links

Gene Details

Gene
RHBDD2
Gene Name
rhomboid domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040456.1 619 Silent Mutation ATC,ATT I192I NP_001035546.1
NM_001040457.1 619 Silent Mutation ATC,ATT I51I NP_001035547.1
XM_005250511.4 619 Silent Mutation ATC,ATT I51I XP_005250568.1

View Full Product Details