Product Details

SNP ID
rs117448709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77136118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAAGGTGAAAACAACGTATCAA[C/T]GTTTGTGGAATGCAGTGAAAGCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC146 PubMed Links

Gene Details

Gene
CCDC146
Gene Name
coiled-coil domain containing 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020879.2 Intron NP_065930.2

View Full Product Details