Product Details

SNP ID
rs112104928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:82755555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACATCAGTCTCTGATGACTCA[A/G]TGAAATTGTGGGAAAAACTGAGATG
Phenotype
MIM: 604918
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCLO PubMed Links

Gene Details

Gene
PCLO
Gene Name
piccolo presynaptic cytomatrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014510.2 18738 Intron NP_055325.2
NM_033026.5 18738 UTR 3 NP_149015.2
XM_017012006.1 18738 UTR 3 XP_016867495.1
XM_017012007.1 18738 Intron XP_016867496.1

View Full Product Details