Product Details

SNP ID
rs113592238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87349158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCCTGTTCCTTCACTTGAAGA[A/G]TCATTAAAAAAATACCTTGAATCAG
Phenotype
MIM: 606090 MIM: 616403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CROT PubMed Links

Gene Details

Gene
CROT
Gene Name
carnitine O-octanoyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143935.1 273 Silent Mutation GAA,GAG E30E NP_001137407.1
NM_001243745.1 273 Silent Mutation GAA,GAG E30E NP_001230674.1
NM_021151.3 273 Silent Mutation GAA,GAG E30E NP_066974.2
XM_011516337.2 273 Silent Mutation GAA,GAG E30E XP_011514639.1
XM_017012370.1 273 UTR 5 XP_016867859.1
Gene
TP53TG1
Gene Name
TP53 target 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details