Product Details

SNP ID
rs116158145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94598534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGTTTAATATCAGTGTCAATT[C/T]CTTAAATATTAAGGTATATGTAGTC
Phenotype
MIM: 611686 MIM: 604149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASD1 PubMed Links

Gene Details

Gene
CASD1
Gene Name
CAS1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022900.4 Intron NP_075051.4
XM_006716092.3 Intron XP_006716155.1
XM_006716093.3 Intron XP_006716156.2
XM_011516495.2 Intron XP_011514797.1
XM_017012546.1 Intron XP_016868035.1
XM_017012547.1 Intron XP_016868036.1
Gene
SGCE
Gene Name
sarcoglycan epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099400.1 Intron NP_001092870.1
NM_001099401.1 Intron NP_001092871.1
NM_001301139.1 Intron NP_001288068.1
NM_003919.2 Intron NP_003910.1
XM_005250675.4 Intron XP_005250732.1
XM_005250677.4 Intron XP_005250734.1
XM_011516663.2 Intron XP_011514965.1
XM_011516664.2 Intron XP_011514966.1
XM_011516665.2 Intron XP_011514967.1
XM_011516666.2 Intron XP_011514968.1
XM_011516667.2 Intron XP_011514969.1
XM_011516669.2 Intron XP_011514971.1
XM_017012763.1 Intron XP_016868252.1
XM_017012764.1 Intron XP_016868253.1
XM_017012765.1 Intron XP_016868254.1
XM_017012766.1 Intron XP_016868255.1
XM_017012767.1 Intron XP_016868256.1
XM_017012768.1 Intron XP_016868257.1
XM_017012769.1 Intron XP_016868258.1
XM_017012770.1 Intron XP_016868259.1

View Full Product Details