Product Details

SNP ID
rs113803296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100091908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGGGGAGCAGCCCCTGAGCACC[A/C]CTGCTGGTGGCTCTGTCCTCTGTTA
Phenotype
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 MIM: 194510
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
There are no transcripts associated with this gene.

Gene
COPS6
Gene Name
COP9 signalosome subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006833.4 1134 UTR 3 NP_006824.2
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278595.1 1134 Intron NP_001265524.1
NM_005916.4 1134 Intron NP_005907.3
NM_182776.2 1134 Intron NP_877577.1
XM_005250348.3 1134 Intron XP_005250405.1
XM_017012217.1 1134 Intron XP_016867706.1
Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

Gene
ZNF3
Gene Name
zinc finger protein 3
There are no transcripts associated with this gene.

View Full Product Details