Product Details
- SNP ID
-
rs112886587
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100681758 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCTCGATCTCACCAGAAGATCC[A/G]TGCAGGGAGTACCCTGAAGCCGGGG
- Phenotype
-
MIM: 612064
MIM: 139390
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GIGYF1
PubMed Links
Gene Details
- Gene
- GIGYF1
- Gene Name
- GRB10 interacting GYF protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022574.4 |
3859 |
Silent Mutation |
CAC,CAT |
H1023H |
NP_072096.2 |
XM_005250532.1 |
3859 |
Silent Mutation |
CAC,CAT |
H1023H |
XP_005250589.1 |
XM_011516472.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1029M |
XP_011514774.1 |
XM_011516477.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1028M |
XP_011514779.1 |
XM_017012526.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1054M |
XP_016868015.1 |
XM_017012527.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1054M |
XP_016868016.1 |
XM_017012528.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1054M |
XP_016868017.1 |
XM_017012529.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1054M |
XP_016868018.1 |
XM_017012530.1 |
3859 |
Missense Mutation |
ACG,ATG |
T1054M |
XP_016868019.1 |
XM_017012531.1 |
3859 |
Missense Mutation |
ACG,ATG |
T952M |
XP_016868020.1 |
- Gene
- GNB2
- Gene Name
- G protein subunit beta 2
There are no transcripts associated with this gene.
View Full Product Details