Product Details

SNP ID
rs117256700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128711295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGATAGAGCTTCTATTTCACAG[A/G]AAATGAGTAGCTAGAGAATCAAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM71F1 PubMed Links

Gene Details

Gene
FAM71F1
Gene Name
family with sequence similarity 71 member F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282788.1 Intron NP_001269717.1
NM_001282789.1 Intron NP_001269718.1
NM_032599.3 Intron NP_115988.1
XM_017012743.1 Intron XP_016868232.1
Gene
FAM71F2
Gene Name
family with sequence similarity 71 member F2
There are no transcripts associated with this gene.

View Full Product Details