Product Details

SNP ID
rs115482016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:132793438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAGAAATAATAATTAGAGCTAT[C/T]TAATATCTCTTTACCCTCATGAAGA
Phenotype
MIM: 613748
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD3 PubMed Links

Gene Details

Gene
CHCHD3
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317177.1 Intron NP_001304106.1
NM_001317178.1 Intron NP_001304107.1
NM_017812.3 Intron NP_060282.1

View Full Product Details