Product Details

SNP ID
rs113064737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19243436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCGGCACATTCACAATGTCATG[C/T]AAGTATTACCACAAACTAAATGTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND4C PubMed Links

Gene Details

Gene
DENND4C
Gene Name
DENN domain containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017925.6 Intron NP_060395.5
XM_006716804.3 Intron XP_006716867.1
XM_006716805.3 Intron XP_006716868.1
XM_006716807.3 Intron XP_006716870.1
XM_011517959.2 Intron XP_011516261.1
XM_017014917.1 Intron XP_016870406.1
XM_017014918.1 Intron XP_016870407.1
XM_017014919.1 Intron XP_016870408.1
XM_017014920.1 Intron XP_016870409.1

View Full Product Details