Product Details

SNP ID
rs111033848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34646729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTCGCGCAGTGGAACCGATCCT[C/T]AGCAACGCCAGCAGGCGTCAGAGGC
Phenotype
MIM: 606999 MIM: 600939 MIM: 601978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALT PubMed Links

Gene Details

Gene
GALT
Gene Name
galactose-1-phosphate uridylyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000155.3 141 Nonsense Mutation CAG,TAG Q9* NP_000146.2
NM_001258332.1 141 UTR 5 NP_001245261.1
Gene
IL11RA
Gene Name
interleukin 11 receptor subunit alpha
There are no transcripts associated with this gene.

Gene
SIGMAR1
Gene Name
sigma non-opioid intracellular receptor 1
There are no transcripts associated with this gene.

View Full Product Details