Product Details

SNP ID
rs112202528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35753265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTCACACCCAGCCGGGAAGTC[A/G]ATGGGATGCTGGGACCTGGGGAACC
Phenotype
MIM: 609471 MIM: 612191 MIM: 108961 MIM: 615742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBA2 PubMed Links

Gene Details

Gene
GBA2
Gene Name
glucosylceramidase beta 2
There are no transcripts associated with this gene.

Gene
MSMP
Gene Name
microseminoprotein, prostate associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044264.2 321 Silent Mutation ATC,ATT I85I NP_001037729.1
Gene
NPR2
Gene Name
natriuretic peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003995.3 321 Intron NP_003986.2
XM_005251478.3 321 Intron XP_005251535.1
XM_005251479.4 321 UTR 5 XP_005251536.1
XM_011517890.2 321 UTR 5 XP_011516192.1
XM_011517891.2 321 UTR 5 XP_011516193.1
XM_011517892.2 321 UTR 5 XP_011516194.1
XM_011517893.2 321 UTR 5 XP_011516195.1
XM_011517895.2 321 Intron XP_011516197.1
XM_017014745.1 321 UTR 5 XP_016870234.1
XM_017014746.1 321 UTR 5 XP_016870235.1
XM_017014747.1 321 UTR 5 XP_016870236.1
XM_017014748.1 321 UTR 5 XP_016870237.1
XM_017014749.1 321 UTR 5 XP_016870238.1
XM_017014750.1 321 UTR 5 XP_016870239.1
Gene
RGP1
Gene Name
RGP1 homolog, RAB6A GEF complex partner 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080496.2 321 UTR 3 NP_001073965.2

View Full Product Details