Product Details

SNP ID
rs117928572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35864318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTAAATCTAATTTTAACCACTT[A/G]ATACTTATTTTAGGCTTCACAAATT
Phenotype
MIM: 616888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00950 PubMed Links

Gene Details

Gene
LINC00950
Gene Name
long intergenic non-protein coding RNA 950
There are no transcripts associated with this gene.

Gene
OR13J1
Gene Name
olfactory receptor family 13 subfamily J member 1
There are no transcripts associated with this gene.

Gene
TMEM8B
Gene Name
transmembrane protein 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042589.2 11882 Intron NP_001036054.1
NM_001042590.2 11882 Intron NP_001036055.1
NM_016446.3 11882 Intron NP_057530.2
XM_005251483.4 11882 Intron XP_005251540.3
XM_011517901.2 11882 Intron XP_011516203.1
XM_011517902.2 11882 Intron XP_011516204.1
XM_011517903.2 11882 Intron XP_011516205.1
XM_011517904.2 11882 Intron XP_011516206.1
XM_011517905.1 11882 Intron XP_011516207.1
XM_011517908.2 11882 Intron XP_011516210.1
XM_011517910.2 11882 Intron XP_011516212.1
XM_011517911.2 11882 Intron XP_011516213.1
XM_011517912.2 11882 Intron XP_011516214.1
XM_011517913.2 11882 Intron XP_011516215.1
XM_011517914.2 11882 Intron XP_011516216.1
XM_011517915.2 11882 Intron XP_011516217.1
XM_011517916.2 11882 UTR 3 XP_011516218.1
XM_011517918.2 11882 Intron XP_011516220.1
XM_017014805.1 11882 Intron XP_016870294.1
XM_017014806.1 11882 Intron XP_016870295.1
XM_017014807.1 11882 Intron XP_016870296.1

View Full Product Details