Product Details

SNP ID
rs116579314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5510544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTTGACTCACTCTCATGTTAC[A/G]GCAAACCTTAAGCTGAATGAACAAC
Phenotype
MIM: 605723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDCD1LG2 PubMed Links

Gene Details

Gene
PDCD1LG2
Gene Name
programmed cell death 1 ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025239.3 107 Intron NP_079515.2
XM_005251600.3 107 UTR 5 XP_005251657.1

View Full Product Details