Product Details
- SNP ID
-
rs111867781
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:81585623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCTCCATGCCCACTGCCAGCCA[C/T]TCCCCGGTGGGGCAGTACCCCAGGG
- Phenotype
-
MIM: 600189
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLE1
PubMed Links
Gene Details
- Gene
- TLE1
- Gene Name
- transducin like enhancer of split 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001303103.1 |
2246 |
Silent Mutation |
GAA,GAG |
E680E |
NP_001290032.1 |
NM_001303104.1 |
2246 |
Silent Mutation |
GAA,GAG |
E655E |
NP_001290033.1 |
NM_005077.4 |
2246 |
Silent Mutation |
GAA,GAG |
E670E |
NP_005068.2 |
XM_005252151.1 |
2246 |
Silent Mutation |
GAA,GAG |
E714E |
XP_005252208.1 |
XM_005252152.1 |
2246 |
Silent Mutation |
GAA,GAG |
E713E |
XP_005252209.1 |
XM_005252153.1 |
2246 |
Silent Mutation |
GAA,GAG |
E704E |
XP_005252210.1 |
XM_005252154.1 |
2246 |
Silent Mutation |
GAA,GAG |
E703E |
XP_005252211.1 |
XM_005252156.2 |
2246 |
Silent Mutation |
GAA,GAG |
E679E |
XP_005252213.1 |
XM_005252162.1 |
2246 |
Silent Mutation |
GAA,GAG |
E630E |
XP_005252219.1 |
XM_005252163.2 |
2246 |
Silent Mutation |
GAA,GAG |
E596E |
XP_005252220.1 |
XM_006717258.1 |
2246 |
Silent Mutation |
GAA,GAG |
E713E |
XP_006717321.1 |
XM_006717259.3 |
2246 |
Silent Mutation |
GAA,GAG |
E658E |
XP_006717322.1 |
XM_006717260.1 |
2246 |
Silent Mutation |
GAA,GAG |
E640E |
XP_006717323.1 |
XM_006717261.2 |
2246 |
Silent Mutation |
GAA,GAG |
E599E |
XP_006717324.1 |
XM_006717262.1 |
2246 |
Silent Mutation |
GAA,GAG |
E669E |
XP_006717325.1 |
XM_006717263.1 |
2246 |
Silent Mutation |
GAA,GAG |
E668E |
XP_006717326.1 |
XM_011518951.2 |
2246 |
Silent Mutation |
GAA,GAG |
E669E |
XP_011517253.1 |
XM_017015064.1 |
2246 |
Silent Mutation |
GAA,GAG |
E679E |
XP_016870553.1 |
XM_017015065.1 |
2246 |
Silent Mutation |
GAA,GAG |
E678E |
XP_016870554.1 |
XM_017015066.1 |
2246 |
Silent Mutation |
GAA,GAG |
E624E |
XP_016870555.1 |
View Full Product Details