Product Details

SNP ID
rs112353960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86026288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTGAGGCTGGAAGAGGACTTA[G/T]AAGAGTATGAAAGTACTCTAAGATT
Phenotype
MIM: 606804
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GOLM1 PubMed Links

Gene Details

Gene
GOLM1
Gene Name
golgi membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016548.3 2933 UTR 3 NP_057632.2
NM_177937.2 2933 UTR 3 NP_808800.1
Gene
NAA35
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321881.1 2933 Intron NP_001308810.1
NM_001321882.1 2933 Intron NP_001308811.1
NM_024635.3 2933 Intron NP_078911.3
XM_005252127.3 2933 Intron XP_005252184.1
XM_011518903.2 2933 UTR 3 XP_011517205.1
XM_011518904.2 2933 UTR 3 XP_011517206.1
XM_017015012.1 2933 UTR 3 XP_016870501.1

View Full Product Details