Product Details

SNP ID
rs113783037
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.9:104694610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCTTTTTCTGTTCCTGCTCTG[C/T]CTCATCATGTACATGATAATCCTCC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 189 Silent Mutation TGC,TGT C63C NP_001004484.1

View Full Product Details