Product Details

SNP ID
rs117219732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:112754960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTAGACGTCCTCTGAAGTCTTC[A/G]AAATAGAGAAGGCTTTTCCTCCATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNX30 PubMed Links

Gene Details

Gene
SNX30
Gene Name
sorting nexin family member 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012994.1 Intron NP_001013012.1
XM_011518691.2 Intron XP_011516993.1
XM_017014716.1 Intron XP_016870205.1

View Full Product Details