Product Details

SNP ID
rs114036737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125442353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTTGCAGAGAAGCATCTGCTT[C/T]GAGGGAGCATTTGGACCTGTAAAAT
Phenotype
MIM: 610558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPKAP1 PubMed Links

Gene Details

Gene
MAPKAP1
Gene Name
mitogen-activated protein kinase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006617.1 Intron NP_001006618.1
NM_001006618.1 Intron NP_001006619.1
NM_001006619.1 Intron NP_001006620.1
NM_001006620.1 Intron NP_001006621.1
NM_001006621.1 Intron NP_001006622.1
NM_024117.3 Intron NP_077022.1
XM_011519005.1 Intron XP_011517307.1
XM_011519006.2 Intron XP_011517308.1
XM_011519007.1 Intron XP_011517309.1
XM_011519008.1 Intron XP_011517310.1
XM_011519009.1 Intron XP_011517311.1
XM_011519010.2 Intron XP_011517312.1
XM_017015126.1 Intron XP_016870615.1
XM_017015127.1 Intron XP_016870616.1

View Full Product Details