Product Details

SNP ID
rs111686018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128721411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACGAGGAGCAGGCTGGCCACCA[A/G]CGAGAAGAGGGACAGCAGCAGGAAG
Phenotype
MIM: 610714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 762 Intron NP_001304855.1
NM_013355.4 762 Intron NP_037487.2
XM_005251946.3 762 Intron XP_005252003.1
XM_006717080.2 762 Intron XP_006717143.1
XM_017014649.1 762 Intron XP_016870138.1
XM_017014650.1 762 Intron XP_016870139.1
Gene
ZDHHC12
Gene Name
zinc finger DHHC-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318015.1 762 Silent Mutation CTG,TTG L247L NP_001304944.1
NM_001318016.1 762 Missense Mutation GCT,GTT A201V NP_001304945.1
NM_001318020.1 762 Silent Mutation CGC,CGT R184R NP_001304949.1
NM_001318023.1 762 Silent Mutation CTG,TTG L191L NP_001304952.1
NM_032799.4 762 Silent Mutation CTG,TTG L192L NP_116188.2
XM_011519116.2 762 Silent Mutation CGC,CGT R239R XP_011517418.1
Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
There are no transcripts associated with this gene.

View Full Product Details