Product Details

SNP ID
rs112933307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18113101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCTCCTGACTGTCCAAGGCAG[G/T]CAAGCTCTGCTCACTCATTCCTGGC
Phenotype
MIM: 607229
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRGPRX3 PubMed Links

Gene Details

Gene
MRGPRX3
Gene Name
MAS related GPR family member X3
There are no transcripts associated with this gene.

Gene
SAA3P
Gene Name
serum amyloid A3 pseudogene
There are no transcripts associated with this gene.

Gene
SAAL1
Gene Name
serum amyloid A like 1
There are no transcripts associated with this gene.

View Full Product Details