Product Details

SNP ID
rs111613383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18173775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTGGTGCTGATTCTAGTTGGTG[C/T]GAAACGTCAGATTTCATCCCAGTCG
Phenotype
MIM: 607230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX4 PubMed Links

Gene Details

Gene
MRGPRX4
Gene Name
MAS related GPR family member X4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054032.3 Intron NP_473373.2

View Full Product Details