Product Details

SNP ID
rs117051022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18707039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGCCCCGTCACTGTGTACTTG[C/T]TGCTGAAGACACGCTCGGCTGCCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173588.3 4463 Missense Mutation AAC,AGC N1152S NP_775859.3
XM_011520030.2 4463 Missense Mutation AAC,AGC N1172S XP_011518332.1
XM_011520031.2 4463 Missense Mutation AAC,AGC N1071S XP_011518333.1
XM_017017607.1 4463 Missense Mutation AAC,AGC N1172S XP_016873096.1
XM_017017608.1 4463 Missense Mutation AAC,AGC N1152S XP_016873097.1
Gene
TMEM86A
Gene Name
transmembrane protein 86A
There are no transcripts associated with this gene.

View Full Product Details