Product Details

SNP ID
rs117009146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19055225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGGCTGTGGTGGAAGCCTGTGA[A/G]TAAGGCAGTTCCAGAGACACTGCAC
Phenotype
MIM: 607228
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX2 PubMed Links

Gene Details

Gene
MRGPRX2
Gene Name
MAS related GPR family member X2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303615.1 1266 UTR 3 NP_001290544.1
NM_054030.3 1266 UTR 3 NP_473371.1

View Full Product Details