Product Details

SNP ID
rs115354234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19055309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCACAAGGACTCTCTTAACTG[C/T]TTTAAAATCAGGACTGAGAAAGTTC
Phenotype
MIM: 607228
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX2 PubMed Links

Gene Details

Gene
MRGPRX2
Gene Name
MAS related GPR family member X2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303615.1 1182 UTR 3 NP_001290544.1
NM_054030.3 1182 UTR 3 NP_473371.1

View Full Product Details