Product Details

SNP ID
rs117029992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2268840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGTGCCCTTAGAAAATCCTCT[C/T]CCCTCCGGAAAAATCTCCAAGTGTT
Phenotype
MIM: 601886
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASCL2 PubMed Links

Gene Details

Gene
ASCL2
Gene Name
achaete-scute family bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005170.2 1507 UTR 3 NP_005161.1

View Full Product Details