Product Details

SNP ID
rs112835699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGCCATTGTGGGGAATATGACC[A/G]TTCTCTTTGTGATCAAAACTGAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 139 Missense Mutation ATT,GTT I47V NP_001005165.1

View Full Product Details