Product Details

SNP ID
rs111493577
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTATCCTAGCTTCTGTGGTTG[C/T]TGGAAGAAATTTAGTTCTTGTAACC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 452 Missense Mutation GCT,GTT A151V NP_001005165.1

View Full Product Details