Product Details

SNP ID
rs117363131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6236419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAACAAGCTGATAAGGCCCAAA[C/T]TAGAGTAGCAAAAATGCAGAGAAGT
Phenotype
MIM: 609472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf42 PubMed Links

Gene Details

Gene
C11orf42
Gene Name
chromosome 11 open reading frame 42
There are no transcripts associated with this gene.

Gene
CNGA4
Gene Name
cyclic nucleotide gated channel alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037329.3 Intron NP_001032406.1
XM_017017217.1 Intron XP_016872706.1
XM_017017218.1 Intron XP_016872707.1
XM_017017219.1 Intron XP_016872708.1
Gene
FAM160A2
Gene Name
family with sequence similarity 160 member A2
There are no transcripts associated with this gene.

View Full Product Details